Please login to your online account to display your discounted pricing

Sodium Chromate, 4-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$162.04 - $327.42

Chemical Identifiers

Chemical Name or Material Sodium Chromate
CAS 10034-82-9
Name Note 4-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Na2CrO4·4H2O
Formula Weight 234.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004051

 
avantor performance
364004
Poly Bottle 125g Each for $162.04

02004052

 
avantor performance
364001
Poly Bottle 500g Each for $327.42
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 234.03
Chemical Name or Material Sodium Chromate
Name Note 4-Hydrate
Packaging Poly Bottle
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Na2CrO4·4H2O
CAS 10034-82-9
Quantity 125g