Learn More
Sodium chlorodifluoroacetate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1575622
Description
It is used as pharmaceutical intermediate. Reactions of aldehydes with triphenylphosphine and sodium chlorodifluoroacetate yield 1,1-difluoro olefins. It is a general reaction and a one step synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediate. Reactions of aldehydes with triphenylphosphine and sodium chlorodifluoroacetate yield 1,1-difluoro olefins. It is a general reaction and a one step synthesis.
Solubility
Soluble in dimethoxyethane, diglyme (0.60 g mL-1), triglyme, dimethyl formamide.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Sodium chlorodifluoroacetate | |
∼195°C (decomposition) | |
MFCD00064771 | |
4162363 | |
Soluble in dimethoxyethane,diglyme (0.60 g mL-1),triglyme,dimethyl formamide. | |
C(=O)(C(F)(F)Cl)[O-].[Na+] | |
152.457 | |
152.46 |
1895-39-2 | |
C2ClF2NaO2 | |
100 g | |
sodium chlorodifluoroacetate, sodium 2-chloro-2,2-difluoroacetate, chlorodifluoroacetic acid sodium salt, sodium chloro difluoro acetate, acetic acid, chlorodifluoro-, sodium salt, sodiumchlorodifluoroacetate, difluorochloroacetic acid sodium salt, sodium difluorochloroacetate, chlorodifluoroacetic acid sodium | |
MRTAVLDNYYEJHK-UHFFFAOYSA-M | |
sodium;2-chloro-2,2-difluoroacetate | |
2734985 | |
97% |
Chemical Identifiers
1895-39-2 | |
152.457 | |
MRTAVLDNYYEJHK-UHFFFAOYSA-M | |
2734985 | |
C(=O)(C(F)(F)Cl)[O-].[Na+] |
C2ClF2NaO2 | |
MFCD00064771 | |
sodium chlorodifluoroacetate, sodium 2-chloro-2,2-difluoroacetate, chlorodifluoroacetic acid sodium salt, sodium chloro difluoro acetate, acetic acid, chlorodifluoro-, sodium salt, sodiumchlorodifluoroacetate, difluorochloroacetic acid sodium salt, sodium difluorochloroacetate, chlorodifluoroacetic acid sodium | |
sodium;2-chloro-2,2-difluoroacetate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 217-586-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only