Please login to your online account to display your discounted pricing

Sodium Chlorite, Technical Grade, 80%, LabChem™

$218.51 - $218.51

Chemical Identifiers

Chemical Name or Material Sodium Chlorite
CAS 7758-19-2
CAS Max % 80
CAS 7647-14-5
CAS Max % 20
Purity Grade Technical Grade
Linear Formula NaClO2
Formula Weight 90.44
UN Number UN1496
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC236201

 
lab chem
LC236201
500g Each for $218.51
Description & Specifications

Specifications

Formula Weight 90.44
Solubility Soluble in water
Color White
Decomposition Information Carbon monoxide; Carbon dioxide; Hydrogen chloride; Sodium oxide
Health Hazard 1 Danger
Physical Form Crystals
CAS Max % 80
Health Hazard 2 GHS H Statement
May intensify fire; oxidizer.
Toxic if swallowed.
Fatal in contact with skin or if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
CAS Max % 20
Linear Formula NaClO2
Health Hazard 3 GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe dust.
Do not get in eyes, on skin, or on clothing.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear respiratory protection.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Collect spillage.
Store in a well-ventilated place.
Keep container tightly closed.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Assay 80%
Packaging Poly Bottle
UN Number UN1496
Chemical Name or Material Sodium Chlorite
Identification Passes Test
Purity Grade Technical Grade
CAS 7758-19-2
Storage Information Room Temperature
CAS 7647-14-5
Quantity 500g
Safety and Handling

Danger

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe dust.
Do not get in eyes, on skin, or on clothing.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear respiratory protection.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Collect spillage.
Store in a well-ventilated place.
Keep container tightly closed.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May intensify fire; oxidizer.
Toxic if swallowed.
Fatal in contact with skin or if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.