missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium chlorite, Tech. nominally 80%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1426536

 View more versions of this product

Catalog No. AA1426536



Specifications

Specifications

Sodium chlorite
Nominally 80%
MFCD00003478
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
YZHUMGUJCQRKBT-UHFFFAOYSA-M
sodium;chlorate
516902
90.44
Technical
500g
7775-09-9,7775-09-9,1310-73-2,497-19-8,7647-14-5,7757-82-6
ClNaO3
White flake/powder
Sodium chlorite: 80.0%; Sodium chlorate: 3.0%; Sodium hydroxide: 3.0%; Sodium carbonate: 2.0%; Sodium chloride: 10.0%; Sodium sulphate: 0.3%; Water: 1.7%
[O-]Cl(=O)=O.[Na+]
106.44
CHEBI:65242
Flakes or Powder
White

Chemical Identifiers

7775-09-9
106.44
YZHUMGUJCQRKBT-UHFFFAOYSA-M
516902
sodium;chlorate
ClNaO3
MFCD00003478
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
CHEBI:65242
[O-]Cl(=O)=O.[Na+]
Safety and Handling

Safety and Handling

H272-H301-H310-H314-H412: May intensify fire; oxidizer. Toxic if swallowed. Fatal in contact with skin. Causes severe skin burns and eye damage. Harmful to aquatic life with long lasting effects.

DOTInformation : Hazard Class: 5.1, Packing Group: II

EINECSNumber : 231-836-6

Documents