Learn More
Sodium chlorite, 80%, pure, unstabilized, Thermo Scientific Chemicals
Sodium chlorite, 80%, ClNaO2, CAS Number-7758-19-2, 7647-14-5, unii-g538ebv4vf, alcide ld, neo silox d, chlorous acid, sodium salt, caswell no. 755, sodiumchlorite, textone, sodium chlorite, textile, chlorite sodium, 500g, CHEBI:78667, White, 231-836-6, 05,603; 09,423; 11,481; 13,280, 90.44, Pure
$84.33 - $460.90
Chemical Identifiers
CAS | 7758-19-2 |
---|---|
MDL Number | MFCD00003478 |
InChI Key | UKLNMMHNWFDKNT-UHFFFAOYSA-M |
Synonym | sodium chlorite, chlorous acid, sodium salt, textone, chlorite sodium, textile, alcide ld, neo silox d, caswell no. 755, sodiumchlorite, unii-g538ebv4vf |
PubChem CID | 23668197 |
ChEBI | CHEBI:78667 |
IUPAC Name | sodium;chlorite |
SMILES | [O-]Cl=O.[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC223230050
|
Thermo Scientific Chemicals
223230050 |
5 g | Glass Bottle |
Each for $84.33
|
|
||||
AC223235000
|
Thermo Scientific Chemicals
223235000 |
500 g | Glass Bottle |
Each for $140.21
|
|
||||
AC223230025
|
Thermo Scientific Chemicals
223230025 |
2.5 kg | Plastic Bottle |
Each for $460.90
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-19-2 | |
UKLNMMHNWFDKNT-UHFFFAOYSA-M | |
23668197 | |
sodium;chlorite |
MFCD00003478 | |
sodium chlorite, chlorous acid, sodium salt, textone, chlorite sodium, textile, alcide ld, neo silox d, caswell no. 755, sodiumchlorite, unii-g538ebv4vf | |
CHEBI:78667 | |
[O-]Cl=O.[Na+] |
Specifications
180.0°C to 200.0°C | |
White | |
5 g | |
ClNaO2 | |
MFCD00003478 | |
15, 8735 | |
Solubility in water: 390g/L (17°C). | |
[O-]Cl=O.[Na+] | |
90.44 | |
CHEBI:78667 | |
80% | |
Glass Bottle |
7758-19-2,7647-14-5 | |
Powder | |
75% min. (Iodometry) | |
NaClO2 | |
05,603; 09,423; 11,481; 13,280 | |
sodium chlorite, chlorous acid, sodium salt, textone, chlorite sodium, textile, alcide ld, neo silox d, caswell no. 755, sodiumchlorite, unii-g538ebv4vf | |
UKLNMMHNWFDKNT-UHFFFAOYSA-M | |
sodium;chlorite | |
23668197 | |
90.44 | |
Pure | |
Sodium chlorite, Unstabilized |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Toxic if swallowed.
Fatal if inhaled.
May intensify fire; oxidizer.
Contact with acids liberates very toxic gas.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date
Danger
EINECSNumber : 231-836-6
RTECSNumber : VZ4800000
TSCA : TSCA
RUO – Research Use Only