missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Chloride USP/FCC Granular, Macron Fine Chemicals™

macron

$685.60 - $685.60

Chemical Identifiers

CAS 7647-14-5
Physical Form Granular
Molecular Formula NaCl
Formula Weight 58.44
Chemical Name or Material Sodium Chloride
Grade FCC/USP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK7532212 Avantor Performance
753206
Poly 2.5kg Each for $685.60
Case of 6 Each for $4,113.60
Specifications Description & Specifications

Chemical Identifiers

7647-14-5
NaCl
Sodium Chloride
Granular
58.44
FCC/USP

Specifications

7647-14-5
NaCl
Poly
Sodium Chloride
Granular
58.44
2.5kg
FCC/USP