missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Chloride RV, EP (Euopean Pharmacopoeia) primary standard, Ricca Chemical, 50g

$962.97 - $1,269.67

Chemical Identifiers

CAS 7647-14-5
Physical Form Crystals
Chemical Name or Material Sodium Chloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
71-922-50 SDS Ricca Chemical Company
RI7192250
250g Each for $1,269.67
72-278-50G Ricca Chemical Company
RI7227.850G
50g Each for $962.97
Specifications Description & Specifications

Chemical Identifiers

7647-14-5
Crystals
Sodium Chloride

Specifications

250g
Colorless, white
Sodium Chloride
Crystals
7647-14-5