Please login to your online account to display your discounted pricing

Sodium chloride, pure, ca. 26% solution in water, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC387640010

 View more versions of this product

Catalog No. AC387640010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 58.44
Quantity 1L
Color Clear colorless
Purity Grade Pure
Density 1.2020g/mL
Linear Formula NaCl
CAS 7732-18-5
Chemical Name or Material Sodium chloride
MDL Number MFCD00003477
CAS 7647-14-5
Molecular Formula ClNa
Health Hazard 1
Packaging Glass bottle
Concentration 25 to 28%

Specifications

Chemical Name or Material Sodium chloride
CAS 7647-14-5
CAS 7732-18-5
Purity Grade Pure
Molecular Formula ClNa
Linear Formula NaCl
Formula Weight 58.44
MDL Number MFCD00003477
Accessories

SDS & Safety Documents