missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium chloride, pure, ca. 26% solution in water, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 387640025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Sodium chloride | |
2.5L | |
NaCl | |
15,8734 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium;chloride | |
5234 | |
58.44 | |
Glass bottle | |
1.2020g/mL |
7647-14-5,7732-18-5 | |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
Pure | |
1.202 |
RUO â Research Use Only