missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium chloride, Optical Grade

Manufacturer:  Alfa Aesar™ 1101918

 View more versions of this product

Catalog No. AA1101918



Specifications

Specifications

Sodium chloride
ClNa
14,8599
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium;chloride
5234
58.44
Optical
Odorless
Hygroscopic
White
6 to 9
7647-14-5
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
[Na+].[Cl-]
58.44
CHEBI:26710
Broken Crystals
2.165g/mL
1.5442
1413°C
801°C
50g

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
sodium;chloride
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710
[Na+].[Cl-]
Safety and Handling

Safety and Handling

RTECSNumber : VZ4725000

TSCA : TSCA

Documents