missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega Sodium Chloride, Molecular Biology Grade, Promega™ DFS Item

Used in many molecular biology and forensic applications.

Manufacturer:  Promega H5273

 View more versions of this product

Catalog No. PRH5273



Description

Description

  • Quality tested and certified free of DNase, RNase and protease activity
Specifications

Specifications

Sodium Chloride
≥99.5%
common salt, dendritis, halite, purex, rock salt, saline, salt, sodium chloride, sodium chloride nacl, table salt
[Na+].[Cl-]
58.44
CHEBI:26710
Solid
1kg
7647-14-5,7732-18-5
ClNa
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
5234
58.45
Molecular Biology

Chemical Identifiers

ClNa
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
sodium chloride
58.44
common salt, dendritis, halite, purex, rock salt, saline, salt, sodium chloride, sodium chloride nacl, table salt
CHEBI:26710
[Na+].[Cl-]
Safety and Handling

Safety and Handling

Recommended Storage : Store at +15°C to +30°C

Documents