Learn More
Sodium Chloride, For Sulfate (Contains Hydrochloric Acid), 240g/L, LabChem™
Supplier: LabChem 235101
Specifications
Sodium Chloride | |
For Sulfate (Contains Hydrochloric Acid) | |
20.79,78.33,0.88 | |
NaCl | |
UN1789 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
5234 | |
58.44 | |
Poly Bottle | |
500 mL |
Colorless | |
7647-14-5,7732-18-5,7647-01-0 | |
ClNa | |
MFCD00003477 | |
240g/L | |
Soluble in water | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
Passes Test | |
Hydrogen chloride; Thermal decomposition generates: Corrosive vapors | |
Liquid |
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
5234 | |
sodium chloride |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
CHEBI:26710 | |
[Na+].[Cl-] |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature