Please login to your online account to display your discounted pricing

Sodium Chloride, For Sulfate (Contains Hydrochloric Acid), 240g/L, LabChem™

$118.59 - $118.59

Chemical Identifiers

Chemical Name or Material Sodium Chloride
CAS 7732-18-5
CAS Max % 78.33
CAS 7647-14-5
CAS Max % 20.79
CAS 7647-01-0
CAS Max % 0.88
Name Note For Sulfate (Contains Hydrochloric Acid)
Linear Formula NaCl
Formula Weight 58.44
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC235101

 
lab chem
235101
500mL Each for $118.59
Description & Specifications

Specifications

Formula Weight 58.44
Name Note For Sulfate (Contains Hydrochloric Acid)
CAS 7732-18-5
Solubility Soluble in water
Chemical Name or Material Sodium Chloride
CAS 7647-14-5
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
Quantity 500mL
Identification Passes Test
CAS Max % 78.33
Physical Form Liquid
Linear Formula NaCl
CAS Max % 20.79
Storage Information Room Temperature
Health Hazard 1 Danger
Color Clear, colorless
Decomposition Information Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
UN Number UN1789
CAS 7647-01-0
Concentration 240g/L
CAS Max % 0.88
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.