Please login to your online account to display your discounted pricing

Sodium Chloride, For Chloride, Certified, 0.0250N ±0.0005N (0.025M), LabChem™

$63.23 - $63.23

Chemical Identifiers

Chemical Name or Material Sodium Chloride
CAS 7732-18-5
CAS Max % 99.85
CAS 7647-14-5
CAS Max % 0.15
Name Note For Chloride
Purity Grade Certified Grade
Linear Formula NaCl
Formula Weight 58.44
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC235501

 
lab chem
LC235501
500mL Each for $63.23
Description & Specifications

Specifications

Formula Weight 58.44
Purity Grade Certified Grade
CAS Max % 0.15
Color Clear, colorless
Linear Formula NaCl
CAS 7647-14-5
Quantity 500mL
Identification Passes Test
Chemical Name or Material Sodium Chloride
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 99.85
Physical Form Liquid
Concentration 0.0250N ±0.0005N (0.025M)
Storage Information Room Temperature
Traceability to NIST Traceable to NIST
Name Note For Chloride
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Solubility Soluble in water
CAS 7732-18-5
Decomposition Information Hydrogen chloride
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.