missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium chloride, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 194090050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1461.0°C | |
801.0°C | |
White | |
Crystalline Powder | |
ClNa | |
MFCD00003477 | |
15, 8734 | |
Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol (1g/10mL), ammonia, slightly soluble in alcohol, nearly insoluble in hydrochloric acid | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
≥99.0% | |
Plastic Bottle |
Sodium chloride | |
7647-14-5 | |
5.0 to 8.0 (5%, H<2O, at 20°C) | |
5 kg | |
NaCl | |
13,280 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
5234 | |
58.44 | |
Extra Pure |
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
5234 | |
[Na+].[Cl-] |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
CHEBI:26710 |
Safety and Handling
EINECSNumber : 231-598-3
RUO – Research Use Only