missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium chloride, crystalline powder PDV, 99+%

Manufacturer:  Alfa Aesar™ A123130I

 View more versions of this product

Catalog No. AAA123130I



Description

Description

Applications
Sodium chloride is widely used as a food additive, and a preservative. It is employed in the Chloralkali process for producing chlorine and sodium hydroxide, in the Solvay process for producing sodium carbonate and calcium chloride, in the Mannheim process and in the Hargreaves process for producing sodium sulfate and hydrochloric acid. It is used in the manufacture of sodium chlorate, water softening and de-icing of roadways. Further it is used in several industries like pharmaceutical, textile, dyeing, rubber food and agriculture. It is used in the processing of steel, copper, vanadium, and other metals.

Solubility
Freely soluble in water. Slightly soluble in alcohol and ammonia.

Notes
It is sensitive to moisture in air. It should be protected in an air tight container.
Specifications

Specifications

Sodium chloride
ClNa
14,8599
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium;chloride
5234
58.44
≥99%
1413°C
6 to 9
7647-14-5
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
[Na+].[Cl-]
58.44
CHEBI:26710
Crystalline Powder Pdv
Odorless
801°C
5000g

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
sodium;chloride
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710
[Na+].[Cl-]
Safety and Handling

Safety and Handling

RTECSNumber : VZ4725000

TSCA : TSCA

SDS Documents