Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium chloride crystal optic rectangle, 38.5mm x 19.5mm x 4mm, polished both sides

Chemical Identifiers

Chemical Name or Material Sodium chloride crystal optic rectangle
CAS 7647-14-5
Name Note 38.5mm x 19.5mm x 4mm, Polished both sides
MDL Number MFCD00003477
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38467KT

 
alfa aesar™
38467KT
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sodium chloride crystal optic rectangle
TSCA TSCA
Odor Odorless
Name Note 38.5mm x 19.5mm x 4mm, Polished both sides
MDL Number MFCD00003477
CAS 7647-14-5
Quantity 1 each
RTECS Number SC7500000