Please login to your online account to display your discounted pricing

Sodium chloride crystal optic disc, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Sodium chloride crystal
Name Note Polished on both sides
Physical Form Disc, 13mm X 2mm
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38413KT

 
alfa aesar
38413KT
1 piece CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Disc, 13mm X 2mm
Quantity 1 piece
Chemical Name or Material Sodium chloride crystal
Name Note Polished on both sides