Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium chloride crystal optic disc, 13mm x 1mm, polished both sides

Spectroscopy, IR window

Chemical Identifiers

Chemical Name or Material Sodium chloride crystal optic disc
Name Note 13mm x 1mm, Polished both sides
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38443KT

  SDS
alfa aesar™
38443KT
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Name Note 13mm x 1mm, Polished both sides
Odor Odorless
Color White
Quantity 1 each
Chemical Name or Material Sodium chloride crystal optic disc
TSCA TSCA