Please login to your online account to display your discounted pricing

Sodium Chloride, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$97.11 - $146.61

Chemical Identifiers

Chemical Name or Material Sodium Chloride
CAS 7647-14-5
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula NaCl
Formula Weight 58.44
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004043

 
avantor performance
362419
Poly Bottle 1kg Each for $146.61

02004047

 
avantor performance
362401
Poly Bottle 500g Each for $97.11
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 58.44
Chemical Name or Material Sodium Chloride
CAS 7647-14-5
Molecular Formula NaCl
Specific Gravity 1L = 2.16kg
Packaging Poly Bottle
Quantity 1kg