missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Chloride CP Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7647-14-5
Physical Form Granular
Molecular Formula NaCl
Formula Weight 58.44
Chemical Name or Material Sodium Chloride
Grade CP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK7544212 Avantor Performance
754406
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7647-14-5
NaCl
Sodium Chloride
Granular
58.44
CP

Specifications

58.44
Granular
NaCl
Sodium Chloride
2.5kg
7647-14-5
Poly
CP