missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Chloride (colorless to white crystalline granules/USP/FCC/EP/BP/JP), Fisher Chemical

This product is not intended for dialysis, parenteral or injectable use without further processing

$165.21 - $749.00

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium;chloride
SMILES [Na+].[Cl-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
S64212 SDS View Documents Thermo Scientific
S64212
12kg Poly Pail Each for $749.00
 
S642212 SDS View Documents Thermo Scientific
S642212
2.5kg Poly Bottle Each for $243.50
Case of 4 Each for $974.00
 
S642500 SDS View Documents Thermo Scientific
S642500
500g Poly Bottle Each for $165.21
 
Specifications

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
sodium;chloride
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710
[Na+].[Cl-]

Specifications

7647-14-5
Pass Test (BP, JP, EP, USP), Pass Test (BP, JP, EP, Pass Test (EP), Pass Test (JP), Pass Test (USP)
≤100ppm (EP), 100ppm max. (EP)
ClNa
Pass Test (BP), Pass Test (BP, EP, USP), Pass Test (EP), Pass Test (USP)
≤1ppm (BP, EP, USP), ≤1mg/kg (FCC), ≤2ppm (JP), 1mg/kg max. (FCC), 1ppm max. (BP), 1ppm max. (EP), 1ppm max. (USP), 2ppm max. (JP)
[Na+].[Cl-]
sodium;chloride
58.44
CHEBI:26710
Solid
BP/EP/FCC/JP/USP
≤100ppm (BP), ≤0.010% (Limit of) (USP), 0.010% max. (USP), 100ppm max. (BP)
Pass Test (JP)
2.165g/cm³
Pass Test (BP, EP, JP, USP), Pass Test (BP, EP, JP, Pass Test (EP), Pass Test (JP), Pass Test (USP)
2mg/kg max. (FCC), 5ppm max. (USP), 5ppm max. (USP), 2mg/kg max. (FCC)
Pass Test (BP, EP, JP, USP), Pass Test (BP, EP, JP, Pass Test (EP), Pass Test (JP), Pass Test (USP)
≤0.5% (at 110°C, FCC), ≤0.5% (BP/BP, EP/EP, JP/JP, USP/USP), 0.5% max. (BP), 0.5% max. (EP), 0.5% max. (FCC), 0.5% max. (JP), 0.5% max. (USP)
Poly Pail
0.05% max. (USP), 0.05% max. (USP), 500ppm max. (EP/BP), 500ppm max. (BP), 500ppm max. (EP)
≤0.02% (JP), ≤0.020% (USP), ≤200ppm (BP, EP), 0.02% max. (JP), 0.020% max. (USP), 200ppm max. (BP), 200ppm max. (EP)
1461°C
801°C
12kg
Vapor Pressure: 1mmHg at 440°C
≤100ppm (BP), ≤0.01% (JP, USP), 0.01% max. (JP), 0.01% max. (USP), 100ppm max. (BP)
0.2ppm max. (BP), 0.2ppm max. (EP), 0.2ppm max. (USP), 0.2ppm max. (USP/EP/BP)
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5 I.U./g max. (EP)
Pass Test (BP, EP, JP, USP), Pass Test (BP, EP, JP, Pass Test (EP), Pass Test (JP), Pass Test (USP)
5234
58.44
99.0 to 100.5% (EP)
≤100ppm (EP), ≤0.01% (JP), 0.01% max. (JP), 100ppm max. (EP)
≤0.35% (FCC), 0.35% max. (FCC)
Pass Test (JP)
5 I.U./g max. (BP), 5 I.U./g max. (EP)
≤5ppm (BP, EP), ≤3ppm (JP), 3ppm max. (JP), 5ppm max. (BP), 5ppm max. (EP)
Pass Test (BP, SP, FCC, JP, USP), Pass Test (BP, SP, FCC, Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (USP)
≤2ppm (BP, EP, USP), ≤2μg/g (JP), 2μg/g max. (JP), 2ppm max. (BP), 2ppm max. (EP), 2ppm max. (USP)
Pass Test (BP), Pass Test (BP, EP, USP), Pass Test (EP), Pass Test (USP)
≤25ppm (BP, EP), ≤0.0025% (JP, USP), 0.0025% max. (JP), 0.0025% max. (USP), 25ppm max. (BP), 25ppm max. (EP)
Meets Requirements (BP), Meets Requirements (BP, EP, USP), Meets Requirements (EP), Meets Requirements (USP)
1mmHg at 865°C
White
5.0 to 8.0
Sodium Chloride
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1