missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Chloride (colorless to white crystalline granules/USP/FCC/EP/BP), Fisher Chemical™
$275.38 - $6222.77
Chemical Identifiers
CAS | 7647-14-5 |
---|---|
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
IUPAC Name | sodium chloride |
SMILES | [Na+].[Cl-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
S641-500
|
Fisher Chemical
FLS641500 |
500 g | Poly Bottle |
Each for $275.38
Case of 6 Each for $1,652.28
|
|
||||
S641-350LB
|
Fisher Chemical
FLS641350LB |
350 lb. | Fiber Drum |
Each for $6,222.77
|
|
||||
S641-212
|
Fisher Chemical
FLS641212 |
2.5 kg | Poly Bottle |
Each for $670.38
Case of 4 Each for $2,681.52
|
|
||||
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
5234 | |
sodium chloride |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
CHEBI:26710 | |
[Na+].[Cl-] |
Specifications
1461°C | |
7647-14-5 | |
5.0 to 8.0 | |
500 g | |
MFCD00003477 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
5234 | |
58.44 | |
BP/EP/FCC/USP | |
0.5% max. (BP), 0.5% max. (EP), 0.5% max. (FCC), 0.5% max. (USP), 0.5% max. (USP/FCC/EP/BP) | |
1mmHg at 865°C | |
Sodium Chloride |
801°C | |
White | |
Solid | |
ClNa | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
99.0 to 100.5% (EP) | |
Pass Test (BP), Pass Test (EP), Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC/EP/BP) | |
Poly Bottle | |
2.165g/cm³ |