Please login to your online account to display your discounted pricing

Sodium Chloride, Certified, 9.0% (w/v) ±0.1%, LabChem™

$64.71 - $64.71

Chemical Identifiers

Chemical Name or Material Sodium Chloride
CAS 7732-18-5
CAS Max % 91.5
CAS 7647-14-5
CAS Max % 8.5
Purity Grade Certified Grade
Linear Formula NaCl
Formula Weight 58.44
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC234854

 
lab chem
LC234854
4L Each for $64.71
Description & Specifications

Specifications

Formula Weight 58.44
Assay 9.0% ± 0.1% (w/v)
Purity Grade Certified Grade
Solubility Soluble in water
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Identification Passes Test
Specific Gravity 1.06g/mL
Chemical Name or Material Sodium Chloride
Quantity 4L
Density 1.06g/mL
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Linear Formula NaCl
Packaging Poly Bottle
Decomposition Information Hydrogen chloride
CAS Max % 91.5
Storage Information Room Temperature
Physical Form Liquid
CAS 7732-18-5
CAS 7647-14-5
CAS Max % 8.5
Color Clear, colorless
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.