Please login to your online account to display your discounted pricing

Sodium Chloride, Certified, 1.000N ±0.005N (0.1M), LabChem™

$76.46 - $76.46

Chemical Identifiers

Chemical Name or Material Sodium Chloride
CAS 7732-18-5
CAS Max % 94.4
CAS 7647-14-5
CAS Max % 5.6
Purity Grade Certified Grade
Linear Formula NaCl
Formula Weight 58.44
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC235802

 
lab chem
LC235802
1L Each for $76.46
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 58.44
CAS Max % 94.4
Identification Passes Test
Quantity 1L
CAS 7732-18-5
Density 1.04g/mL
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Concentration 1.000N ±0.005N (0.1M)
Color Clear, colorless
CAS Max % 5.6
Packaging Poly Bottle
Linear Formula NaCl
CAS 7647-14-5
Storage Information Room Temperature
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Physical Form Liquid
Chemical Name or Material Sodium Chloride
Solubility Soluble in water
Specific Gravity 1.04g/mL
Decomposition Information Hydrogen chloride
Traceability to NIST Traceable to NIST
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.