missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Chloride, ACS Grade, ≥ 99.0%, LabChem™

$60.49 - $60.49

Chemical Identifiers

CAS 7647-14-5
CAS Max % 100
Linear Formula NaCl
Formula Weight 58.44
Chemical Name or Material Sodium Chloride
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC234201 Lab Chem
LC234201
500g Each for $60.49
Specifications Description & Specifications

Chemical Identifiers

7647-14-5
NaCl
Sodium Chloride
100
58.44
ACS Grade

Specifications

7647-14-5
Crystals
58.44
Passes Test
Soluble in water
Room Temperature
1,413°C
801°C
Sodium Chloride
ACS Grade
100
NaCl
2.165g/cm3
Poly Bottle
2.165g/cm3
Hydrogen chloride
White
500g
≥99.0%

Safety and Handling

GHS H Statement
Product is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

EINECSNumber : 231-598-3