Please login to your online account to display your discounted pricing

Sodium Chloride ACS, EMD Millipore


Traceable To NIST

Quantity 12kg
Packaging Poly Pail

Manufacturer: emd millipore corporation  SX04205

 View more versions of this product

Catalog No. MSX04205

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Grade
Formula Weight 58.44
Quantity 12kg
Bulk Density 1140kg/m3
Density 2.17g/cm3 (20°C)
Name Note Certipure™, Secondary Reference Material For Argentometry
CAS 7647-14-5
Vapor Pressure 1.3 hPa (865°C)
Packaging Poly Pail
Chemical Name or Material Sodium Chloride
Molecular Formula ClNa
Recommended Storage Store at +15°C to +25°C
Linear Formula NaCl

Specifications

Chemical Name or Material Sodium Chloride
Name Note Certipure™, Secondary Reference Material For Argentometry
CAS 7647-14-5
Purity Grade ACS Grade
Molecular Formula ClNa
Linear Formula NaCl
Formula Weight 58.44
Accessories

Store at +15°C to +25°C