Please login to your online account to display your discounted pricing

Sodium Chloride ACS, EMD Millipore

Traceable To NIST

$35.43 - $839.74

Chemical Identifiers

Chemical Name or Material Sodium Chloride
Name Note Certipure™, Secondary Reference Material For Argentometry
CAS 7647-14-5
Purity Grade ACS Grade
Molecular Formula ClNa
Linear Formula NaCl
Formula Weight 58.44
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MSX04203

 
emd millipore corporation
SX04203
Poly Bottle 2.5kg Each for $102.29

MSX04205

 
emd millipore corporation
SX04205
Poly Pail 12kg Each for $207.69

MSX04201

 
emd millipore corporation
SX04201
Poly bottle 500g Each for $35.43

MSX042022

 
emd millipore corporation
SX042022
Fiber Drum w/bag 50kg Each for $839.74
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 58.44
Quantity 2.5kg
Bulk Density 1140kg/m3
Density 2.17g/cm3 (20°C)
Chemical Name or Material Sodium Chloride
Name Note Certipure™, Secondary Reference Material For Argentometry
Packaging Poly Bottle
Molecular Formula ClNa
Linear Formula NaCl
CAS 7647-14-5
Vapor Pressure 1.3 hPa (865°C)
Recommended Storage Store at +15°C to +25°C
Safety and Handling

Store at +15°C to +25°C