Please login to your online account to display your discounted pricing

Sodium Chloride ACS AR Crystal, Macron Fine Chemicals™


macron

Quantity 12kg
Packaging Poly Pail

Manufacturer: avantor performance  758119

 View more versions of this product

Catalog No. MK7581P12

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Poly Pail
Quantity 12kg
Chemical Name or Material Sodium Chloride
CAS 7647-14-5
Purity Grade ACS
Physical Form Crystalline
Molecular Formula NaCl
Formula Weight 58.44g/mol

Specifications

Chemical Name or Material Sodium Chloride
CAS 7647-14-5
Purity Grade ACS
Physical Form Crystalline
Molecular Formula NaCl
Formula Weight 58.44g/mol