missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Chloride, ACS, 99.5% min, MilliporeSigma™

$64.18 - $124.74

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1064040500
View Documents Promotion Details
Technidata
1064040500
500 g Poly Bottle
Each for $64.18
 
M1064041000
View Documents Promotion Details
Technidata
1064041000
1 kg Plastic Bottle
Each for $124.74
 
Description

Description

Specifications

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
sodium chloride
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710
[Na+].[Cl-]

Specifications

Colorless
Crystalline
99.5% min.
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
5234
58.44
1.3 hPa (865°C)
Sodium Chloride, For ACS analysis
7647-14-5
500 g
ClNa
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
[Na+].[Cl-]
58.44
CHEBI:26710
Poly Bottle
2.17 g/cm3 (20°C)
SDS
missing translation for 'documents'

missing translation for 'documents'