missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium chloride, ACS, 99.0% min., Alfa Aesar™

Manufacturer:  Alfa Aesar 1231436

 View more versions of this product

Catalog No. AA1231436



Specifications

Specifications

Sodium chloride
7647-14-5
ClNa
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
[Na+].[Cl-]
58.44
CHEBI:26710
Crystalline
500g
For analysis ACS
99.0% min
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium;chloride
5234
58.44
2.165g/mL

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
sodium;chloride
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
CHEBI:26710
[Na+].[Cl-]
Safety and Handling

Safety and Handling

EINECSNumber : 231-598-3

Documents