Please login to your online account to display your discounted pricing

Sodium chloride, 99.99%, Suprapur™, EMD Millipore


Quantity 50g
Packaging Plastic Bottle

Manufacturer: emd millipore corporation  1.06406.0050

 View more versions of this product

Catalog No. M1064060050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solid
Formula Weight 58.44
Color Colorless
Quantity 50g
Density 2.17g/cm3 (20°C)
Purity Grade Suprapur
Solubility 358g/L (20°C)
Molecular Formula NaCl
Assay Percent Range 99.99%
Linear Formula NaCl
Name Note Sodium chloride
Vapor Pressure 1.3 hPa (865°C)
Packaging Plastic Bottle
Additional Information Vapor Pressure: 1.3 hPa (865°C)
CAS 7647-14-5

Specifications

Name Note Sodium chloride
CAS 7647-14-5
Purity Grade Suprapur
Physical Form Solid
Assay Percent Range 99.99%
Molecular Formula NaCl
Linear Formula NaCl
Formula Weight 58.44