Please login to your online account to display your discounted pricing

Sodium Chloride, 99.0% min., EMD Millipore


Manufacturer: emd millipore corporation  SX04253

 View more versions of this product

Catalog No. MSX04253

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals
Formula Weight 58.44
Color White
Quantity 2.5kg
Chemical Name or Material Sodium Chloride
Synonym Rock salt
Assay Percent Range 99.9% min.
CAS 7647-14-5
Molecular Formula NaCl

Specifications

Chemical Name or Material Sodium Chloride
CAS 7647-14-5
Assay Percent Range 99.9% min.
Molecular Formula NaCl
Synonym Rock salt