Please login to your online account to display your discounted pricing

Sodium Chloride, 99.0% min., EMD Millipore

$207.37 - $207.37

Chemical Identifiers

Chemical Name or Material Sodium Chloride
CAS 7647-14-5
Assay Percent Range 99.9% min.
Molecular Formula NaCl
Synonym Rock salt
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

MSX04253

 
emd millipore corporation
SX04253
Each for $207.37
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 58.44
Color White
Quantity 2.5kg
Chemical Name or Material Sodium Chloride
Assay Percent Range 99.9% min.
Molecular Formula NaCl
CAS 7647-14-5
Synonym Rock salt