Please login to your online account to display your discounted pricing

Sodium Chloride, 9% (w/v) Aqueous Solution, Ricca Chemical

$44.09 - $70.85

Chemical Identifiers

Chemical Name or Material Sodium Chloride
CAS 7732-18-5
CAS Min % 90.0
CAS Max % 94.0
CAS 7647-14-5
CAS Min % 8.1
CAS Max % 8.43
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

7215-91

 
ricca chemical company
RI7215.91
Poly natural 4L Each for $70.85

7215-932

 
ricca chemical company
RI7215.932
Poly natural 1L Each for $44.09

7215-92-5

 
ricca chemical company
RI7215.92.5
Cubitainer 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

7215-95

 
ricca chemical company
RI7215.95
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 4L
Physical Form Liquid
Color Clear colorless
CAS Min % 90.0
CAS Max % 94.0
Chemical Name or Material Sodium Chloride
Shelf Life 24 months
Packaging Poly natural
CAS 7732-18-5
Concentration or Composition (by Analyte or Components) 9% (w/v) Aqueous Solution
CAS 7647-14-5
CAS Min % 8.1
CAS Max % 8.43
Safety and Handling

24 months