missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium chloride, 5M aq. soln., RNAse free

$87.55 - $87.55

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym common salt, dendritis, halite, purex, rock salt, saline, salt, sodium chloride, sodium chloride nacl, table salt
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium;chloride
SMILES [Na+].[Cl-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60434AK View Documents Alfa Aesar™
J60434AK
250mL Each for $87.55
 
Description

Description

Applications
Sodium Chloride is widely used in biochemistry and molecular biology applications. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media. a component of PBS and SSC buffers. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.

Solubility
Soluble in water (358 mg/ml at 20°C), alcohol (very slightly), glycerol (100 mg/ml), and ammonia. Insoluble in hydrochloric acid.

Notes
Incompatible with strong oxidizing agents. Store in a cool, dry, well-ventilated area away from incompatible substances. Store protected from moisture.
Specifications

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
5234
sodium;chloride
ClNa
MFCD00003477
common salt, dendritis, halite, purex, rock salt, saline, salt, sodium chloride, sodium chloride nacl, table salt
CHEBI:26710
[Na+].[Cl-]

Specifications

7647-14-5
NaCl
common salt, dendritis, halite, purex, rock salt, saline, salt, sodium chloride, sodium chloride nacl, table salt
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium;chloride
5234
58.44
5M aq. soln.
Sodium chloride, RNAse free
ClNa
MFCD00003477
Soluble in water (358mg/ml at 20°C),alcohol (very slightly),glycerol (100mg/ml),and ammonia. Insoluble in hydrochloric acid.
[Na+].[Cl-]
58.44
CHEBI:26710
Liquid
250mL
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures