Please login to your online account to display your discounted pricing

Sodium Chloride, 25% (w/v) Aqueous Solution, APHA for Barium, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

72195-1

 
ricca chemical company
RI7219.51
Plastic 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

72195-32

 
ricca chemical company
RI7219.532
Plastic 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

72195-16

 
ricca chemical company
RI7219.516
Plastic 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 4L
Chemical Name or Material Sodium Chloride
Concentration or Composition (by Analyte or Components) 25% (w/v) Aqueous Solution
Methods APHA for Barium
Packaging Plastic