Please login to your online account to display your discounted pricing

Sodium Chloride, 20% (w/v) Aqueous Solution, APHA for Iodide, Ricca Chemical

$52.24 - $90.85

Chemical Identifiers

Chemical Name or Material Sodium Chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

72186-16

  SDS
ricca chemical company
RI7218.616
Plastic 500mL Each for $52.24

72186-32

 
ricca chemical company
RI7218.632
Plastic 1L Each for $90.85
Description & Specifications

Specifications

Quantity 500mL
Methods APHA for Iodide
Packaging Plastic
Concentration or Composition (by Analyte or Components) 20% (w/v) Aqueous Solution
Chemical Name or Material Sodium Chloride