missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium chloride, 0.1N Standardized Solution, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 035616K2
Description
Sodium chloride, 0.1N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium chloride | |
1 L | |
ClNa | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
5234 | |
58.44 | |
23 hPa (17mm Hg) at 20°C | |
Liquid |
7647-14-5 | |
Colorless | |
MFCD00003477 | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
Sodium chloride: 0.6%; Water: 99.4% | |
Miscible with water, alcohol and ammonia |
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
CHEBI:26710 |
ClNa | |
MFCD00003477 | |
5234 | |
[Na+].[Cl-] |
Safety and Handling
EINECSNumber : 231-598-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only