missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium chlorate, ACS, 99.0% min, Alfa Aesar™

Maximum level of impurities: Insoluble matter 0.005%, BrO3 0.015%, Cl 0.005%, SO4 0.001%, Heavy metals (as Pb) 0.001%, Fe 5ppm, Ca 0.002%, Mg 0.002%, K 0.01%

Manufacturer:  Thermo Fisher Scientific 4440822

 View more versions of this product

Catalog No. AA4440822

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Sodium chlorate
7775-09-9
ClNaO3
asex
YZHUMGUJCQRKBT-UHFFFAOYSA-M
sodium;chlorate
516902
106.45
ACS
Odorless
248°C
For analysis ACS
99% min.
MFCD00003479
Soluble in water,glycerol,hydrazine,acetone and methanol. Slightly soluble in ethanol and ammonia.
[O-]Cl(=O)=O.[Na+]
106.44
CHEBI:65242
Crystalline
2.5 g/cm3 at 20°C
Colorless
100g

Chemical Identifiers

7775-09-9
106.44
YZHUMGUJCQRKBT-UHFFFAOYSA-M
516902
sodium;chlorate
ClNaO3
MFCD00003479
asex
CHEBI:65242
[O-]Cl(=O)=O.[Na+]
Safety and Handling

Safety and Handling

P210-P220-P221-P260-P264b-P270-P280-P283-P301+P312-P306+P360-P314-P330-P370+P378q-P371+P380+P375-P501c

H271-H302-H373

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 231-887-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents