missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium chlorate, 99% min, Alfa Aesar™

Manufacturer:  Alfa Aesar 1426722

 View more versions of this product

Catalog No. AA1426722

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Sodium chlorate
99% min.
MFCD00003479
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
[O-]Cl(=O)=O.[Na+]
106.44
CHEBI:65242
Granular
Odorless
100g
7775-09-9
ClNaO3
Colorless granules
YZHUMGUJCQRKBT-UHFFFAOYSA-M
sodium;chlorate
516902
106.45
2.5g/cm3
Colorless

Chemical Identifiers

7775-09-9
106.44
YZHUMGUJCQRKBT-UHFFFAOYSA-M
516902
sodium;chlorate
ClNaO3
MFCD00003479
sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
CHEBI:65242
[O-]Cl(=O)=O.[Na+]
Safety and Handling

Safety and Handling

H271-H302-H411-H401: May cause fire or explosion; strong oxidizer. Harmful if swallowed. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.

DOTInformation : Hazard Class: 5.1, Packing Group: II

EINECSNumber : 231-887-4

Documents