missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Chenodeoxycholate, TCI America™

Manufacturer:  TCI America S0941200MG

 View more versions of this product

Catalog No. S0941200MG



Specifications

Specifications

Sodium Chenodeoxycholate
298°C
2646-38-0
MFCD00065452
WDFRNBJHDMUMBL-PCPSIDHTSA-M
sodium;(4R)-4-[(3R,5S,7R,8R,9R,10S,13R,14R,17S)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
134129466
Crystal-Powder at 20°C
White
200mg
C24H39NaO4
Chenodeoxycholic Acid Sodium Salt, Sodium Chenodesoxycholate, Chenodesoxycholic Acid Sodium Salt, 3alpha,7alpha-Dihydroxy-5beta-cholanic Acid Sodium Salt, 3alpha,7alpha-Dihydroxy-5beta-cholan-24-oic Acid Sodium Salt
CC(CCC(=O)[O-])C1CCC2C1(CCC3C2C(CC4C3(CCC(C4)O)C)O)C.[Na+]
414.562
414.56
≥98.0% (GC)

Chemical Identifiers

2646-38-0
414.562
WDFRNBJHDMUMBL-PCPSIDHTSA-M
134129466
CC(CCC(=O)[O-])C1CCC2C1(CCC3C2C(CC4C3(CCC(C4)O)C)O)C.[Na+]
C24H39NaO4
MFCD00065452
Chenodeoxycholic Acid Sodium Salt, Sodium Chenodesoxycholate, Chenodesoxycholic Acid Sodium Salt, 3alpha,7alpha-Dihydroxy-5beta-cholanic Acid Sodium Salt, 3alpha,7alpha-Dihydroxy-5beta-cholan-24-oic Acid Sodium Salt
sodium;(4R)-4-[(3R,5S,7R,8R,9R,10S,13R,14R,17S)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1427

RTECSNumber : FZ2231000

TSCA : No

Documents