Please login to your online account to display your discounted pricing

Sodium Carbonate, (White, crystalline powder/ACS Reagent Grade), MP Biomedicals

$133.32 - $133.32

Chemical Identifiers

Chemical Name or Material Sodium Carbonate
CAS 497-19-8
Purity Grade ACS Reagent Grade
Physical Form Crystalline Powder
Molecular Formula Na2CO3
Formula Weight 105.99
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19143701

 
mp biomedicals
19143701
1kg Each for $133.32
Description & Specifications

Specifications

Physical Form Crystalline Powder
Purity Grade ACS Reagent Grade
Formula Weight 105.99
Color White
Quantity 1kg
Chemical Name or Material Sodium Carbonate
Molecular Formula Na2CO3
CAS 497-19-8
Recommended Storage Room Temperature