Please login to your online account to display your discounted pricing

Sodium Carbonate, Monohydrate, Crystal, N.F. - F.C.C., J.T.Baker™

$136.77 - $526.65

Chemical Identifiers

Chemical Name or Material Sodium Carbonate
CAS 5968-11-6
Name Note Monohydrate
Purity Grade FCC/NF
Molecular Formula Na2CO3·H2O
Formula Weight 124
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004013

 
avantor performance
360005
Poly Bottle 2.5kg Each for $526.65

02004017

 
avantor performance
360001
Poly Bottle 500g Each for $136.77
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 124
Purity Grade FCC/NF
CAS 5968-11-6
Molecular Formula Na2CO3·H2O
Quantity 2.5kg
Name Note Monohydrate
Packaging Poly Bottle
Specific Gravity 1L = 2.25kg
Chemical Name or Material Sodium Carbonate