Please login to your online account to display your discounted pricing

Sodium Carbonate, Monohydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  359801

 View more versions of this product

Catalog No. 02004014

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Sodium Carbonate
Name Note Monohydrate
CAS 05968-11-6
Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Molecular Formula Na2CO3·H2O
Formula Weight 124
Packaging Poly Bottle
Specific Gravity 1L = 2.25kg
Quantity 500g

Specifications

Chemical Name or Material Sodium Carbonate
CAS 05968-11-6
Name Note Monohydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na2CO3·H2O
Formula Weight 124