Please login to your online account to display your discounted pricing

Sodium Carbonate, Monohydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$282.85 - $432.92

Chemical Identifiers

Chemical Name or Material Sodium Carbonate
CAS 05968-11-6
Name Note Monohydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na2CO3·H2O
Formula Weight 124
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004014

 
avantor performance
359801
Poly Bottle 500g Each for $282.85

02004015

 
avantor performance
359805
Poly Bottle 2.5kg Each for $432.92
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 124
Physical Form Crystals
Chemical Name or Material Sodium Carbonate
Packaging Poly Bottle
Molecular Formula Na2CO3·H2O
CAS 05968-11-6
Quantity 500g
Name Note Monohydrate
Specific Gravity 1L = 2.25kg