Please login to your online account to display your discounted pricing

Sodium carbonate monohydrate, ACS, 99.5% min, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Sodium carbonate monohydrate
CAS 5968-11-6
Purity Grade ACS Grade
Molecular Formula Na2CO3·H2O
Formula Weight 124.01 (105.99 Anhydrous)
MDL Number MFCD00149177
Merck Index 14,8596
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA36485A3

  SDS
alfa aesar
36485A3
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 124.01 (105.99 Anhydrous)
Melting Point 100°C (in 2H2O)
Quantity 2kg
Chemical Name or Material Sodium carbonate monohydrate
TSCA TSCA
Odor Odorless
Physical Form Granular
Assay ≥99.5%
Sensitivity Hygroscopic
Molecular Formula Na2CO3·H2O
MDL Number MFCD00149177
CAS 5968-11-6
Merck Index 14,8596
Color Colorless to White
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H332-H319;
Harmful if inhaled.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P312-P337+P313;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Density 2.250
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P312-P337+P313;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H332-H319;
Harmful if inhaled.
Causes serious eye irritation.