missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium carbonate monohydrate, 99.6%, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 419480010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium carbonate monohydrate | |
5968-11-6 | |
Crystalline Powder | |
99.6% | |
Na2CO3·H2O | |
07,322 | |
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop | |
MQRJBSHKWOFOGF-UHFFFAOYSA-L | |
disodium;carbonate;hydrate | |
2735133 | |
99.6% | |
13.0% to 15.0% (150°C) |
851.0°C | |
White | |
1 kg | |
CNa2O3·H2O | |
MFCD00149177 | |
15, 8731 | |
Solubility in water: 210g/L. Other solubilities: soluble in 1.8 parts boiling water, in 7 parts glyceral, insoluble in alcohol | |
C(=O)([O-])[O-].O.[Na+].[Na+] | |
124.01 | |
124.01 | |
ACS Reagent | |
Plastic bottle |
Chemical Identifiers
5968-11-6 | |
124.01 | |
MQRJBSHKWOFOGF-UHFFFAOYSA-L | |
2735133 | |
C(=O)([O-])[O-].O.[Na+].[Na+] |
CNa2O3·H2O | |
MFCD00149177 | |
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop | |
disodium;carbonate;hydrate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO – Research Use Only