missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium carbonate monohydrate, 85% min

Manufacturer:  Alfa Aesar™ 14108A1

 View more versions of this product

Catalog No. AA14108A1



Specifications

Specifications

Sodium carbonate monohydrate
CH2Na2O4
14,8596
Soluble in water; Insoluble in alcohol
C(=O)([O-])[O-].O.[Na+].[Na+]
124.00
124.01 (105.99 Anhydrous)
≤85%
Odorless
Colorless to White
1kg
5968-11-6
MFCD00149177
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
MQRJBSHKWOFOGF-UHFFFAOYSA-L
disodium;carbonate;hydrate
2735133
Crystalline
2.250g/mL
Hygroscopic
100°C (in -H2O)

Chemical Identifiers

5968-11-6
124.00
MQRJBSHKWOFOGF-UHFFFAOYSA-L
2735133
C(=O)([O-])[O-].O.[Na+].[Na+]
CH2Na2O4
MFCD00149177
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
disodium;carbonate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H332-H319
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P312-P337+P313
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA

Documents