Learn More
Alfa Aesar™ Sodium carbonate monohydrate, 85% min
Manufacturer: Alfa Aesar™ 14108A7
Specifications
Sodium carbonate monohydrate | |
CH2Na2O4 | |
14,8596 | |
Soluble in water; Insoluble in alcohol | |
C(=O)([O-])[O-].O.[Na+].[Na+] | |
124.00 | |
124.01 (105.99 Anhydrous) | |
≤85% | |
Odorless | |
Colorless to White | |
5kg |
5968-11-6 | |
MFCD00149177 | |
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop | |
MQRJBSHKWOFOGF-UHFFFAOYSA-L | |
disodium;carbonate;hydrate | |
2735133 | |
Crystalline | |
2.250g/mL | |
Hygroscopic | |
100°C (in -H2O) |
Chemical Identifiers
5968-11-6 | |
124.00 | |
MQRJBSHKWOFOGF-UHFFFAOYSA-L | |
2735133 | |
C(=O)([O-])[O-].O.[Na+].[Na+] |
CH2Na2O4 | |
MFCD00149177 | |
sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop | |
disodium;carbonate;hydrate |
Safety and Handling
GHS H Statement
H332-H319
Harmful if inhaled.
Causes serious eye irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P312-P337+P313
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Warning
TSCA : TSCA